Learn More
Invitrogen™ iBright™ Prestained Protein Ladder
For visualization and protein sizing during Electrophoresis and confirmation of Western blot detection.
Brand: Invitrogen™ LC5615
237.12 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Invitrogen iBright Prestained Protein Ladder provide direct visualization during electrophoresis and transfer of Western blots, and chemiluminescent or fluorescent Western blot visualization of two unstained proteins for confirmation of western blot detection.The iBright Prestained Protein Ladder contains twelve recombinant proteins, ten (11 to 250 kDa) that are blue-stained and fluor-labeled for direct and near-IR fluorescent visualization and protein sizing, and two proteins (30 kDa and 80 kDa) that are unstained and contain IgG binding sites to bind the primary and secondary antibodies used for chemiluminescent and fluorescent detection of the target protein. These two unstained proteins are compatible with chemiluminescent substrates and fluorescent antibodies.
- Broad size range: twelve proteins spanning 11 to 250 kDa
- Fluorescent visualization: detect the ten stained proteins using the 670nm red laser or 700nm channel of a fluorescence imager
- Western blot confirmation: detect the two unstained proteins using the detection system for the target protein
- Ready to use: supplied in a loading buffer for direct loading on gels
- Sharp bands: clear, sharp bands for direct visualization on both gel and membrane
- Reference band: 55 kDa band has greater intensity for easy orientation
- Quality tested: each lot evaluated by SDS-PAGE and western blotting
Services
1 year

Specifications
Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol Upon receipt, store at –20°C. Product is shipped on ice. |
|
12 | |
Yes | |
1 color: Blue | |
250, 130, 95, 80, 70, 55, 43, 34, 30, 26, 15, 11 kDa | |
2 x 250 μL | |
Western Blotting, SDS-PAGE |
Colorimetric, NIR Fluorescence, and User defined detection system (2 bands) | |
Protein Ladder | |
11 to 250 kDa | |
Novex™ Midi Gels, Novex™ Mini Gels, Novex™Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Gels, NuPAGE™ MES Gels, NuPAGE™ MOPS Gels, NuPAGE™ Tris-Acetate Gels, Precise™ Tris-Glycine Gels, SDS-PAGE Gels, Precise™ Tris-HEPES Gels | |
iBright™ | |
Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.