Learn More
IDF-11774, MedChemExpress
MedChemExpress IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
Brand:  Medchem Express HY-111387
Encompass_Preferred
 View more versions of this product
Product Code. 16421159
  • 97.20 € / 1mg
Explore more special offers
Description
Description
IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
Specifications
Specifications
IDF-11774
Off-White
C23H32N2O2
DMSO : 60 mg/mL (162.82 mM; Need ultrasonic and warming)
368.51
95.37%
Solid
1429054-28-3
Research
1 mg
CN1CCN(C(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)CC1
368.51
Research
Cancer-Kinase/protease