missing translation for 'onlineSavingsMsg'
Learn More

IDF-11774, MedChemExpress

MedChemExpress IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.

93.15 € - 982.80 €

Specifications

Chemical Name or Material IDF-11774
CAS 1429054-28-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H32N2O2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16484438
Encompass_Preferred
View Documents
Medchem Express
HY-111387
10 mg
318.60 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16430757
Encompass_Preferred
View Documents
Medchem Express
HY-111387
50 mg
982.80 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16427167
Encompass_Preferred
View Documents
Medchem Express
HY-111387
5 mg
186.30 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421159
Encompass_Preferred
View Documents
Medchem Express
HY-111387
1 mg
93.15 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
Specifications

Specifications

IDF-11774
Off-White
C23H32N2O2
CN1CCN(C(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)CC1
368.51
Research
Cancer-Kinase/protease
1429054-28-3
Research
DMSO : 60 mg/mL (162.82 mM; Need ultrasonic and warming)
368.51
95.37%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only