missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 sulfatase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A3F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66112-1-IG-150UL
This item is not returnable.
View return policy
Description
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Specifications
Iduronate 2 sulfatase | |
Monoclonal | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22304, Q08890 | |
IDS | |
Iduronate 2 sulfatase Fusion Protein Ag19095 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1A3F9 | |
Unconjugated | |
IDS | |
IDS, iduronate 2 sulfatase, Idursulfase | |
Mouse | |
Protein G | |
RUO | |
100512880, 15931, 3423, 363513 | |
-20°C | |
Liquid |