missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 sulfatase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A3F9, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | Iduronate 2 sulfatase |
---|---|
Clone | 1A3F9 |
Concentration | 0.52 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16817695
|
Proteintech
66112-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16807695
|
Proteintech
66112-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Specifications
Iduronate 2 sulfatase | |
0.52 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IDS, iduronate 2 sulfatase, Idursulfase | |
IDS | |
IgG1 | |
Protein G | |
Antibody |
1A3F9 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Porcine, Human, Mouse, Rat | |
P22304, Q08890 | |
100512880, 15931, 3423, 363513 | |
Iduronate 2 sulfatase Fusion Protein Ag19095 | |
Primary | |
-20°C | |
IDS |