missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iduronate 2 sulfatase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A3F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66112-1-IG-20UL
This item is not returnable.
View return policy
Description
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Specifications
| Iduronate 2 sulfatase | |
| Monoclonal | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22304, Q08890 | |
| IDS | |
| Iduronate 2 sulfatase Fusion Protein Ag19095 | |
| 20 μL | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1A3F9 | |
| Unconjugated | |
| IDS | |
| IDS, iduronate 2 sulfatase, Idursulfase | |
| Mouse | |
| Protein G | |
| RUO | |
| 100512880, 15931, 3423, 363513 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction