missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NICK™ Columns

For rapid purification of labeled DNA (>20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography

Brand:  Cytiva 17-0855-02


 View more versions of this product

Product Code. 10218564

  • 531.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0.15% Kathon™ CG/ICP Biocide
  • Typical recovery: ≥90% of applied DNA
  • Can accommodate sample volumes up to 100μL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography
Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide
Column
50 Columns
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : reaction mass of:5-chloro-2-methyl-4-isothiazolin-3-one and2-methyl-2Hisothiazol- 3-one

SDS
Documents

Documents

Certificates
Special Offers

Special Offers