missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva NICK™ Columns
For rapid purification of labeled DNA (>20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography
242.00 € - 531.00 €
Specifications
For Use With (Application) | Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography |
---|---|
Includes | Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide |
Product Type | Column |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10597065
|
Cytiva
17085501 |
20 Columns |
242.00 €
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10218564
|
Cytiva
17-0855-02 |
50 Columns |
531.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0.15% Kathon™ CG/ICP Biocide
- Typical recovery: ≥90% of applied DNA
- Can accommodate sample volumes up to 100μL
Specifications
Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography | |
Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide | |
Column |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : reaction mass of: 5-chloro2-methyl-4-isothiazolin-3- one ¢ECno. 247-500-7! and2-methyl-2H-isothiaz