missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[1,2-a]pyridine-2-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10600454

Imidazo[1,2-a]pyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10600454
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10600454

Brand: Thermo Scientific Maybridge CC31351DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 64951-08-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD03419462
InChI Key WQLJLPDGSLZYEP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, h-imidazo 1,2-a pyridine-2-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic, 1h-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, technical gradeShow MoreShow Less
PubChem CID 2062168
IUPAC Name imidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C=CC=CC2=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 193.14
Percent Purity 97%
Chemical Name or Material Sodium imidazo[1, 2-a]pyridine-2-carboxylate 0.5 hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[1,2-a]pyridine-2-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.