missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™ Product Code.: 10279322

Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™

Product Code. 10279322
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10279322

Brand: Thermo Scientific Maybridge CC33152CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 859833-15-1
Molecular Formula C8H6Cl2N2O
Molecular Weight (g/mol) 217.049
MDL Number MFCD07772809
InChI Key LHLAJCRPSZDOFF-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1Show MoreShow Less
PubChem CID 18525720
IUPAC Name imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride
SMILES C1=CC2=NC=CN2C=C1C(=O)Cl.Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 250 mg
Formula Weight 217.05
Percent Purity Tech.
Grade Technical
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbonyl chloride hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.