missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, 99%, Thermo Scientific Chemicals

Imidazole, >-98.5%, C3H4N2, CAS Number-288-32-4, 1,3-diaza-2,4-cyclopentadiene, 1,3-diazole, glyoxalin, glyoxaline, imidazol, imidazole, iminazole, imutex, miazole, pyrro b monazole, 500g, 99%, 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow, 01,492; 02,220, 145 deg.C, 68.08

26.13 € - 1473.00 €

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10686312
SDS View Documents
Thermo Scientific Acros
122020050
5 g Glass Bottle
26.13 €
5g
Please to purchase this item. Need a web account? Register with us today!
10284730
SDS View Documents
Thermo Scientific Acros
122021000
100 g Plastic Bottle
41.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
10696502
SDS View Documents
Thermo Scientific Acros
122025000
500 g Plastic Bottle
133.67 €
500g
Please to purchase this item. Need a web account? Register with us today!
10676122
SDS View Documents
Thermo Scientific Acros
122020020
2 kg Plastic Bottle
439.19 €
2kg
Please to purchase this item. Need a web account? Register with us today!
11976251
SDS View Documents
Thermo Scientific Acros
122020100
10 kg Plastic drum
1473.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
1H-imidazole
C3H4N2
MFCD00005183
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
CHEBI:16069
N1C=CN=C1

Specifications

288-32-4
White to Yellow
145°C
98.5% min. (GC)
MFCD00005183
01,492; 02,220
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
68.08
99%
Imidazole
86.0°C to 90.0°C
255.0°C to 256.0°C
Authentic
C3H4N2
23, 45
15, 4953
Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
N1C=CN=C1
68.08
CHEBI:16069
0.003mbar at 20°C
Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-019-2

Product Identifier
  • Imidazole

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.