missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leica Microsystems Immersion Oil for Microscopes
Use Leica™ Immersion Oil with microscope to increase the resolving power and aperture of the objective lens.
Brand: Leica Microsystems 13614800
This item is not returnable.
View return policy
Description
- 10mL bottle
- For Leica Microscopes
Specifications
Immersion Oil | |
10 mL | |
10mL |
1 Pack | |
Type A |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Immersion Oil
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
- MIXTURE LIST-Contains : Benzyl benzoate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction