missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon-NC Membranes, Triton-Free, 0.45 μm

Mixed Cellulose Esters, Surfactant-free

Brand:  Merck HATF08250


 View more versions of this product

Product Code. 10238200

  • 425.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Mixed Cellulose Esters, Surfactant-free

0.45 μm (0.45 micron filter) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free
Non-sterile
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.).
Blotting Membranes
82 mm
White
0.45 μm
60 mL/min x cm2
Plain
Hydrophilic
82 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Immobilon-NC Triton-free 82mm disc 50/pk MCE .45um

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers