missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Immobilon-NC Membranes, Triton-Free, 0.45 μm
Mixed Cellulose Esters, Surfactant-free
425.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Material | Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10166480
|
Merck
HATF08550 |
85 mm |
425.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10238200
|
Merck
HATF08250 |
82 mm |
425.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mixed Cellulose Esters, Surfactant-free
0.45 μm (0.45 micron filter) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.Specifications
Not Autoclavable | |
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free | |
Non-sterile | |
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.). | |
Blotting Membranes |
White | |
0.45 μm | |
60 mL/min x cm2 | |
Plain | |
Hydrophilic |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Immobilon-NC Triton-free 85mm disc
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose