Learn More
Thermo Scientific™ Immuno Breakable Modules in White and Black
Keep wells in place with these breakable modules in white and black, ensuring fluorescence and luminescence assays are easy to handle.
Brand: Thermo Scientific™ 446473
426.98 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Keep wells securely in place with Thermo Scientific™ Breakable Modules in White and Black, ensuring fluorescence and luminescence assays are easy to handle.
Features:
- High-quality polystyrene wells
- Choice of two passive surfaces to enable assay optimization
- C8 strips provide flexibility with sample size and optimal washing performance
- Easy to handle; single well held as easily as strips
- Compatible with common instrumentation
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays.
White:
- Opaque and reflective surface
- Low cross talk and minimal background
Black:
- Opaque and light-absorbing surface
- Minimum cross talk and back-scattered light
Note:
Availability may vary by country.

Specifications
Breakable Strips | |
Yes | |
Fluorescence | |
60 Cases | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
Round, C Bottom | |
60 | |
None | |
Lipids |
Hydrophobic | |
Strip Plate | |
8 x 12 | |
Adsorption | |
Microplate | |
350 μL | |
250 μL | |
PolySorp | |
Fluorescence | |
10 | |
60/Cs. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
181202 | Certificate of Compliance | 03/12/2024 | 446473 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.