Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Brand: Thermo Scientific™ 460348
Description
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
- Polyethylene, stackable frame is marked for lid orientation
Specifications
White | |
Nunc Modules | |
Non-sterile | |
5 | |
60/Cs. |
Frame | |
Polyethylene | |
60 | |
Immuno Standard Modules |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185279 | Certificate of Compliance | 22/03/2025 | 460348 |
184550 | Certificate of Compliance | 22/02/2025 | 460348 |
184520 | Certificate of Compliance | 18/02/2025 | 460348 |
184402 | Certificate of Compliance | 11/02/2025 | 460348 |
183889 | Certificate of Compliance | 11/12/2024 | 460348 |