Learn More
Indium(III) bromide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
171.86 € - 562.80 €
Chemical Identifiers
CAS | 13465-09-3 |
---|---|
Molecular Formula | Br3In |
Molecular Weight (g/mol) | 354.52 |
MDL Number | MFCD00011057 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10130083
|
Thermo Scientific Acros
389030100 |
10 g | Glass bottle |
171.86 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10350614
|
Thermo Scientific Acros
389030500 |
50 g | Glass bottle |
562.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
13465-09-3 | |
Br3In | |
354.52 | |
MFCD00011057 |
Specifications
656.0°C | |
13465-09-3 | |
Crystalline Powder | |
Br3In | |
MFCD00011057 | |
354.52 | |
99.99% | |
Indium(III) bromide |
220.0°C | |
White | |
99.99% | |
InBr3 | |
15, 4997 | |
354.52 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-692-8
- Indium(III) bromide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.