Learn More
Indium(III) n-octoxide, 98%, Thermo Scientific™
33.00 € - 118.00 €
Chemical Identifiers
CAS | 106461-03-4 |
---|---|
MDL Number | MFCD26967910 |
Synonym | indium 3+ ion octanol |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
106461-03-4 | |
MFCD26967910 | |
indium 3+ ion octanol |
Specifications
∼150°C (decomposition) | |
Yellow | |
MFCD26967910 | |
indium 3+ ion octanol | |
98% | |
Indium(III) n-octoxide |
106461-03-4 | |
Solid | |
UN1325 | |
502.48 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen