missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

126.63 € - 480.39 €

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11360979
View Documents
Thermo Scientific Alfa Aesar
010709.09
10 g
126.63 €
10g
Please to purchase this item. Need a web account? Register with us today!
11370979
View Documents
Thermo Scientific Alfa Aesar
010709.18
50 g
480.39 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1312-43-2
277.63
PJXISJQVUVHSOJ-UHFFFAOYSA-N
[O--].[O--].[O--].[In+3].[In+3]
In2O3
MFCD00011060
diindium(3+) trioxidandiide

Specifications

850°C (volat.)
Powder
MFCD00011060
PJXISJQVUVHSOJ-UHFFFAOYSA-N
diindium(3+) trioxidandiide
277.64
Puratronic™
7.18 g/mL
Indium(III) oxide
1312-43-2
In2O3
14,4952
[O--].[O--].[O--].[In+3].[In+3]
277.63
99.997%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-193-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only