missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 11306249

Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11306249
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11306249

Brand: Thermo Scientific Alfa Aesar 044836.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indium(III) sulfide emerged as a promising low-hazard buffer material and is used to improve the properties of solar cells and also replaces the hazardous CdS. It is a mid band gap semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-24-9
Molecular Formula In2S3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011061
InChI Key SIXIBASSFIFHDK-UHFFFAOYSA-N
IUPAC Name diindium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[In+3].[In+3]

Specifications

Melting Point 1050°C
Color Red to Orange
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water and organic solvents.
Formula Weight 325.83
Percent Purity 99.995%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium (III) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN03L043 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number044836.09,044836.18,044836.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.