missing translation for 'onlineSavingsMsg'
Learn More

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals

Preparation of transparent electrical conducting electrodes

142.00 € - 1250.22 €

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349877
SDS View Documents
Thermo Scientific Alfa Aesar
042229.04
2 g N/A N/A N/A
11359877
SDS View Documents
Thermo Scientific Alfa Aesar
042229.09
10 g
576.87 €
10g
Please to purchase this item. Need a web account? Register with us today!
11339877
SDS View Documents
Thermo Scientific Alfa Aesar
042229.14
25 g
1250.22 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the preparation of transparent electrical conducting electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1312-43-2
277.63
PJXISJQVUVHSOJ-UHFFFAOYSA-N
[O--].[O--].[O--].[In+3].[In+3]
In2O3
MFCD00011060
diindium(3+) trioxidandiide

Specifications

1312-43-2
In2O3
UN1993
Soluble in hexane and toluene; Insoluble in alcohols
[O--].[O--].[O--].[In+3].[In+3]
277.63
Indium oxide, Tin doped, polymeric precursor
Viscous Liquid
MFCD00011060
13°C (55°F)
PJXISJQVUVHSOJ-UHFFFAOYSA-N
diindium(3+) trioxidandiide
Oxide ≈20 wt%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336-H372

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.