Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
13.00 € - 635.00 €
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
15565139
|
Fisherbrand™
FB67857 |
1 g |
13.00 €
1 unit |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15565159
|
Fisherbrand™
FB67877 |
50 g |
21.00 €
1 unit |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15565179
|
Fisherbrand™
FB67897 |
2 kg |
286.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15585149
|
Fisherbrand™
FB67869 |
10 g |
16.00 €
1 unit |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
15585169
|
Fisherbrand™
FB67889 |
500 g |
93.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15585189
|
Fisherbrand™
FB67909 |
10 kg |
635.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15545149
|
Fisherbrand™
FB67865 |
5 g |
14.00 €
1 unit |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
15545169
|
Fisherbrand™
FB67885 |
200 g |
33.00 €
1 unit |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
15525179
|
Fisherbrand™
FB67893 |
1 kg |
117.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15525159
|
Fisherbrand™
FB67873 |
20 g |
17.00 €
1 unit |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15505189
|
Fisherbrand™
FB67901 |
5 kg |
348.00 €
1 unit |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
15505169
|
Fisherbrand™
FB67881 |
100 g |
26.00 €
1 unit |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
15505149
|
Fisherbrand™
FB67861 |
2 g |
13.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
- Class M1 stainless steel calibration weight
- In compliance with OIML R111
Specifications
M1 | |
Stainless Steel | |
Cylindrical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1