Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
31.27 € - 630.70 €
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15535179
|
Fisherbrand™
FB67894 |
1 kg |
120.84 €
1 unit |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
15535159
|
Fisherbrand™
FB67874 |
20 g |
34.77 €
1 unit |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15575139
|
Fisherbrand™
FB67858 |
1 g |
31.27 €
1 unit |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
15575159
|
Fisherbrand™
FB67878 |
50 g |
37.05 €
1 unit |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
15575179
|
Fisherbrand™
FB67898 |
2 kg |
173.84 €
1 unit |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15595149
|
Fisherbrand™
FB67870 |
10 g |
34.19 €
1 unit |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
15595169
|
Fisherbrand™
FB67890 |
500 g |
94.98 €
1 unit |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15595189
|
Fisherbrand™
FB67910 |
10 kg |
630.70 €
1 unit |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15555149
|
Fisherbrand™
FB67866 |
5 g |
32.01 €
1 unit |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
15555169
|
Fisherbrand™
FB67886 |
200 g |
47.49 €
1 unit |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
15515189
|
Fisherbrand™
FB67902 |
5 kg |
346.62 €
1 unit |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15515169
|
Fisherbrand™
FB67882 |
100 g |
40.97 €
1 unit |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
15515149
|
Fisherbrand™
FB67862 |
2 g |
31.54 €
1 unit |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
Description
- Class M1 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
Specifications
M1 | |
Stainless Steel | |
Cylindrical |