missing translation for 'onlineSavingsMsg'
Learn More
Indole-3-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11460473

Indole-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11460473
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11460473

Brand: Thermo Scientific Alfa Aesar A12634.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indole-3-carboxylic acid is used as a raw material in synthesis. The structures of the derivatives of indole-3-carboxylic acid were studied using gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy. Reactant for preparation of: Anticancer agents, Derivatives of amino acids and peptides, Serotonin 5-HT4 receptor antagonists, Primary acylureas, Inhibitors of Gli1-mediated transcription in the Hedgehog pathway, Serotonin 5-HT6 antagonists, Very Late Antigen-4 (VLA-4) sntagonists, EphB3 receptor tyrosine kinase inhibitors, Potential Therapeutic Agent for Alzheimer?s Disease, Vinyl ester pseudotripeptide proteasome inhibitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carboxylic acid is used as a raw material in synthesis. The structures of the derivatives of indole-3-carboxylic acid were studied using gas chromatography with high resolution mass spectrometry (GC-HRMS), ultra-high performance liquid chromatography in combination with high resolution tandem mass spectrometry (UHPLC-HRMS), nuclear magnetic resonance spectroscopy (NMR) and Fourier transform infrared spectroscopy. Reactant for preparation of: Anticancer agents, Derivatives of amino acids and peptides, Serotonin 5-HT4 receptor antagonists, Primary acylureas, Inhibitors of Gli1-mediated transcription in the Hedgehog pathway, Serotonin 5-HT6 antagonists, Very Late Antigen-4 (VLA-4) sntagonists, EphB3 receptor tyrosine kinase inhibitors, Potential Therapeutic Agent for Alzheimer′s Disease, Vinyl ester pseudotripeptide proteasome inhibitors

Solubility
Soluble in 95% Ethanol: 50 mg/ml, also soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-50-6
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005624
InChI Key KMAKOBLIOCQGJP-UHFFFAOYSA-N
Synonym indole-3-carboxylic acid, 3-indoleformic acid, 3-carboxyindole, indole-3-carboxylicacid, 3-indolecarboxylic acid, indole-3-carboxylate, 3-indolylcarboxylic acid, beta-indolylcarboxylic acid, indole-3 carboxylic acid, icoShow MoreShow Less
PubChem CID 69867
ChEBI CHEBI:24809
IUPAC Name 1H-indole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)O

Specifications

Melting Point ∼239°C (decomposition)
Quantity 5 g
Beilstein 129435
Solubility Information Soluble in 95% Ethanol: 50mg/ml,also soluble in methanol.
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Indole-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251471 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12634.06,A12634.03,A12634.14
Lot Number10247183 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA12634.06,A12634.03,A12634.14
Lot Number10245801 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA12634.06
Lot Number10225363 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12634.06,A12634.03,A12634.14
Lot Number10228793 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12634.06,A12634.03,A12634.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indole-3-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.