missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl acetate, 97%, Thermo Scientific Chemicals

56.48 € - 563.00 €

Chemical Identifiers

CAS 608-08-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00014561
InChI Key JBOPQACSHPPKEP-UHFFFAOYSA-N
Synonym 3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
PubChem CID 11841
IUPAC Name 1H-indol-3-yl acetate
SMILES CC(=O)OC1=CNC2=CC=CC=C21
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10061750
View Documents
Thermo Scientific Acros
156660010
1 g Glass bottle
56.48 €
1g
Please to purchase this item. Need a web account? Register with us today!
10476800
View Documents
Thermo Scientific Acros
156660050
5 g Glass bottle
178.89 €
5g
Please to purchase this item. Need a web account? Register with us today!
10055853
View Documents
Thermo Scientific Acros
156660250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

608-08-2
175.19
JBOPQACSHPPKEP-UHFFFAOYSA-N
11841
CC(=O)OC1=CNC2=CC=CC=C21
C10H9NO2
MFCD00014561
3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
1H-indol-3-yl acetate

Specifications

608-08-2
Colorless to Lilac
96% min. (GC)
MFCD00014561
3-acetoxyindole, indoxyl acetate, 3-indoxyl acetate, indoxyl-o-acetate, 3-indolyl acetate, indole, 3-acetato, indoxylacetate, 1h-indol-3-ol, 3-acetate, 1h-indol-3-ol, acetate ester, acetic acid, 3-indolyl ester
JBOPQACSHPPKEP-UHFFFAOYSA-N
1H-indol-3-yl acetate
11841
97%
Indoxyl acetate
128.0°C to 131.0°C
Authentic
C10H9NO2
21, 71
Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform
CC(=O)OC1=CNC2=CC=CC=C21
175.19
175.19
Glistening Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-154-2

Product Identifier
  • Indoxyl acetate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.