missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Inhibitor Removal Resin Product Code.: 11319018

Thermo Scientific Chemicals Inhibitor Removal Resin

Product Code. 11319018
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11319018

Brand: Thermo Scientific Chemicals 042489.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-70-7

Specifications

Product Type Inhibitor Removal Resin
Odor Odorless
Molecular Formula C24H27NO5
MDL Number 9003-70-7
Form Crystalline Solid
For Use With (Application) For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
IUPAC Name tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Molecular Weight (g/mol) 409.48
Quantity 100 g
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW29J011 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number042489.22,042489.14,042489.36
Lot NumberW29J010 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number042489.22,042489.14,042489.36
Lot NumberX03I034 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number042489.22,042489.14,042489.36
Lot NumberQ26I026 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number042489.22,042489.14,042489.36
Lot NumberY06E047 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042489.22,042489.14,042489.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Inhibitor Removal Resin >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.