missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Solids pH Electrodes
A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).
Marque: METTLER TOLEDO™ 51344155
506.14 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Meats
- Cheese
- Seafood
- Dough
- Milk and cream
- Butter, yogurt, ice cream
- Fruits and vegetables
- Ready-made meals
- Solid and semi-solid food samples
Accessories & Care
- Open junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
pH Electrode InLab Solids Pro-ISM | |
Open | |
1 to 11 | |
Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | |
Measuring samples with a tough exterior, spear shaped tip has been designed for easy piercing of fruit, cheese, meats, etc. | |
ARGENTHAL | |
InLab™ Solids Pro-ISM | |
<20 sec. | |
32°F to 176°F | |
Spear | |
25 mm | |
6 mm |
Multi-Pin | |
Glass | |
0°C to 80°C | |
Xerolyt Extra Polymer | |
Multi-Pin to BNC/RCA (30281896) | |
LoT Glass | |
NTC 30 kilohm | |
FRISCOLYT-B | |
0°C to 80°C | |
0.98 in. | |
0.23 in. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu