missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Solids pH Electrodes
A low maintenance glass pH electrode with an integrated temperature probe featuring a clog-free, open junction that eliminates the risk of protein fouling. Its spear shaped tip is designed for piercing solid and semi-solid food samples (IP67 rated).
410.00 € - 505.00 €
Specifications
Junction | Open |
---|---|
Material | Glass |
pH Range | 1 to 11 |
Temperature (Metric) Operating | 0°C to 80°C |
Internal Reference | ARGENTHAL |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10766733
|
METTLER TOLEDO™
51343153 |
InLab™ Solids | Combination Electrode | S7 | S7 to BNC (30281915) |
410.00 €
Each |
In Stock
Log in to see stock. |
||
11374213
|
METTLER TOLEDO™
51344155 |
InLab™ Solids Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
505.00 €
Each |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||
11997771
|
METTLER TOLEDO™
51343156 |
InLab™ Solids Go-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC IP67 | Fixed Cable to sensor |
490.00 €
Each |
In Stock
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Meats
- Cheese
- Seafood
- Dough
- Milk and cream
- Butter, yogurt, ice cream
- Fruits and vegetables
- Ready-made meals
- Solid and semi-solid food samples
Accessories & Care
- Open junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Open | |
1 to 11 | |
ARGENTHAL |
Glass | |
0°C to 80°C | |
LoT Glass |