Learn More
Thermo Scientific™ Integrated Gas Module for Varioskan™ LUX
Control O2 and CO2 levels with this optional, integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader.
9350.00 €
Specifications
Software Type | SkanIt Software |
---|---|
CO2 Concentration Range | 0.1 to 15% |
CO2 Concentration Stability | ±0.3% at 5% CO2 |
For Use With (Equipment) | Varioskan LUX |
O2 Concentration Range | 1 to 21% |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15370777
|
Thermo Scientific™
VLGAS001 |
Integrated gas module (factory installed at instrument purchase) |
9350.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader lets you perform cell-based microplate assays in controlled atmosphere. It can simultaneously control CO2 concentrations from 0.1% to 15% and O2 concentrations from 1% to 21%. The module fits neatly inside the instrument and does not take up extra space in the lab.
- Control CO2 levels from 0.1% to 15% and oxygen levels from 1% to 21%, either simultaneously or individually
- Allows users to set gas levels as part of the protocol setup in the intuitive Thermo Scientific™ SkanIt™ software
- Tracks and records gas levels throughout the run for traceability
- Integrated completely inside the instrument – no extra space needed
- Well-suited for a wide range of cell-based applications, including studies involving long-term cell proliferation, induction and detection of hypoxia (such as in cancer research), and growth of anaerobic bacteria
Specifications
SkanIt Software | |
±0.3% at 5% CO2 | |
1 to 21% |
0.1 to 15% | |
Varioskan LUX |
Note: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.