missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Integrated Gas Module for Varioskan™ LUX

Control O2 and CO2 levels with this optional, integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader.

Brand:  Thermo Scientific™ VLGAS001

7975.49 EUR valid until 2025-12-31
Use promo code "25043" to get your promotional price.


 View more versions of this product

Product Code. 15370777

  • 9640.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Integrated gas module for the Thermo Scientific™ Varioskan™ LUX multimode microplate reader lets you perform cell-based microplate assays in controlled atmosphere. It can simultaneously control CO2 concentrations from 0.1% to 15% and O2 concentrations from 1% to 21%. The module fits neatly inside the instrument and does not take up extra space in the lab.

For many cell-based assays, gas control in a microplate reader saves time and labor by enabling simultaneous incubation and measurement, so researchers don't have to shuttle plates between an incubator and a reader to maintain optimal conditions for cell samples. The Varioskan LUX offers an optional integrated gas module, which allows users to set and monitor atmospheric conditions inside the microplate reader.

  • Control CO2 levels from 0.1% to 15% and oxygen levels from 1% to 21%, either simultaneously or individually
  • Allows users to set gas levels as part of the protocol setup in the intuitive Thermo Scientific™ SkanIt™ software
  • Tracks and records gas levels throughout the run for traceability
  • Integrated completely inside the instrument – no extra space needed
  • Well-suited for a wide range of cell-based applications, including studies involving long-term cell proliferation, induction and detection of hypoxia (such as in cancer research), and growth of anaerobic bacteria

The gas control module can be included with the Varioskan LUX reader at time of purchase. Contact your sales representative for more information.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SkanIt Software
±0.3% at 5% CO2
Varioskan LUX
0.1 to 15%
Integrated gas module (factory installed at instrument purchase)
1 to 21%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Integrated Gas Module for Varioskan™ LUX >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Note: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.