Learn More
Diversey™ IntelliCare Dispenser Hybrid
The IntelliCare Hybrid dispensing system can address all hand care needs with a versatile, modern, and durable platform that is compatible with both liquid/gels and foam products.
Brand:  Diversey™ D7524180
 View more versions of this product
Product Code. 17673105
  • 139.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dispenses 1300 mL bottles of hand soap and 1200 mL of alcohol based hand sanitizer
  • Can dispense 0.4 mL or 1.00 mL per push
  • Hybrid dispenser provides an automatic switch from touchless to manual if batteries died before replacement
  • Near empty product alarm and low battery signal
  • Locking dispenser contains a translucent window for product visibility without product tampering
Specifications
Specifications
Hybrid Dispenser (Manual/Automatic)
White
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Diversey™ IntelliCare Dispenser Hybrid > White