missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jaytec Glass™ Soda Glass Milk and Cream Pipettes
Soda Glass Milk and Cream Pipettes are for use with thick liquids.
Brand: Jaytec Glass™ MP.684
This item is not returnable.
View return policy
Description
- Blowout pipettes with wide bore jets for thick liquids
- The last drop in the jet must be blown-out except where shown
- MP.688 (BS.696) NOT to be blown out
Specifications
Soda Lime Glass | |
Delivery of a precise volume in milk/cream testing | |
±0.08 mL | |
5 mL | |
One-mark Blowout Pipette | |
Butyrometer |
Non-sterile | |
1 | |
Clear | |
355 mm | |
Reusable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Jaytec Glass™ Soda Glass Milk and Cream Pipettes > Soda Glass Milk and Cream Pipettes; Capacity: 5 mL; Tolerance: ±0.08 mL; Type: One-mark Blowout Pipette
Spot an opportunity for improvement?Share a Content Correction