Learn More
Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
120.00 € - 201.00 €
Products 14
Product Code Brand Tube Size No. of Places Depth (Metric) Price Quantity & Availability  
Product Code Brand Tube Size No. of Places Depth (Metric) Price Quantity & Availability  
11490908
View Documents
Grant Instruments™
QB-0
None Solid Block None
120.00 €
Each
In Stock
to see stock.
Add to basket
10211643
View Documents
Grant Instruments™
QB-50
50mL 4 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
10781434
View Documents
Grant Instruments™
QB-24
24mm 5 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
12664915
View Documents
Grant Instruments™
QB-17H
17mm 10 75 mm hole depth
201.00 €
Each
In Stock
to see stock.
Add to basket
11420918
View Documents
Grant Instruments™
QB-13
13mm 12 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11430918
View Documents
Grant Instruments™
QB-16
16mm 12 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
15389625
View Documents
Grant Instruments™
QB-E5
16.7 mm 12 53.5 mm hole depth This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11440918
View Documents
Grant Instruments™
QB-18
18mm 12 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11480918
View Documents
Grant Instruments™
QB-E0
0.5mL 24 30 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11400918
View Documents
Grant Instruments™
QB-10
10mm 24 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11490918
View Documents
Grant Instruments™
QB-E1
1.5mL 24 35 mm hole depth
174.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11400928
View Documents
Grant Instruments™
QB-E2
2.0mL 24 35 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11410918
View Documents
Grant Instruments™
QB-12
12mm 24 50 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
11470918
View Documents
Grant Instruments™
QB-H
0.2mL 56 14 mm hole depth
162.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes