Learn More
Invitrogen™ eBioscience™ IC Fixation Buffer
Can be used to fix cells prior to permeabilization with the Permeabilization Buffer (cat. 00-8333) when performing intracellular staining of antigens located in the cytoplasm and secretory pathway, such as cytokines, chemokines and growth factors.
Brand: Invitrogen™ 00-8222-49
Alert:
Contains formaldehyde
Description
- Compatible with surface-staining fluorochrome-conjugated antibodies
- Can be used as a short-term storage buffer for fluorochrome (including tandem dyes)-stained cells for up to 3 days at 2° to 8°C (stored in the dark)

Specifications
125 mL | |
Fixation Reagent | |
eBioscience™ | |
Fixation Buffer |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- eBioscience IC Fixation Buffer
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Paraformaldehyde
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2519395 | Certificate of Analysis | 21/02/2025 | 00-8222-49 |
2498655 | Certificate of Analysis | 21/02/2025 | 00-8222-49 |
2865823 | Certificate of Analysis | 21/02/2025 | 00-8222-49 |
2917438 | Certificate of Analysis | 21/02/2025 | 00-8222-49 |
2801365 | Certificate of Analysis | 21/02/2025 | 00-8222-49 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.