missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Buccal Cell Kit

ChargeSwitch gDNA Buccal Cell Kit maximizes the yield of high quality genomic DNA from buccal samples for rapid buccal swab processing.

Brand:  Invitrogen™ CS11021

232.01 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10097332

  • 348.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes: ChargeSwitch Lysis Buffer (L11)
ChargeSwitch Magnetic Beads
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch Purification Buffer (N6)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Requires : Use of a magnetic accessory.
This item is not returnable. View return policy

Description

Description

  • Sample size 1 buccal swab; Typical performance up to 6 μg
  • Maximum yield sensitivity
  • Rapid single-tube protocol, suitable for automation
  • High-quality DNA for downstream Single Tandem Repeat (STR) analysis
  • ChargeSwitch gDNA Buccal Cell Kit maximizes the yield of high quality genomic DNA from buccal samples for rapid buccal swab processing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buccal Cell
Kit
Buccal Samples
High-throughput Compatible
ChargeSwitch™
50 Preps
Genomic DNA

All components of the ChargeSwitch™ gDNA Buccal Cell Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L11)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N6)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

ChargeSwitch Chemistry
Forensic
50 Preps
gDNA Buccal Cell Kit
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.