missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ gDNA Buccal Cell Kit

ChargeSwitch gDNA Buccal Cell Kit maximizes the yield of high quality genomic DNA from buccal samples for rapid buccal swab processing.

340.00 € - 3350.00 €

Specifications

Cell Type Buccal Cell
Content And Storage

All components of the ChargeSwitch™ gDNA Buccal Cell Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L11)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N6)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Buccal Samples
View More Specs

Includes: ChargeSwitch Lysis Buffer (L11)
ChargeSwitch Magnetic Beads
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch Purification Buffer (N6)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Requires: Use of a magnetic accessory.


Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10097332
View Documents
Invitrogen™
CS11021
50 Preps
340.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10707433
View Documents
Invitrogen™
CS1102110
960 Preps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Sample size 1 buccal swab; Typical performance up to 6 μg
  • Maximum yield sensitivity
  • Rapid single-tube protocol, suitable for automation
  • High-quality DNA for downstream Single Tandem Repeat (STR) analysis
  • ChargeSwitch gDNA Buccal Cell Kit maximizes the yield of high quality genomic DNA from buccal samples for rapid buccal swab processing

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Buccal Cell
Kit
Buccal Samples
High-throughput Compatible
gDNA Buccal Cell Kit
Genomic DNA

All components of the ChargeSwitch™ gDNA Buccal Cell Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L11)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N6)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

ChargeSwitch Chemistry
Forensic
ChargeSwitch™
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.