missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ PCR Clean-Up Kit

ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts.

Brand:  Invitrogen™ CS1200010

 View more versions of this product

Product Code. 10221853

  • 2345.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Requires : Magnetic accessory
This item is not returnable. View return policy

Description

Description

ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts (Figure 1). This kit is scalable for use on liquid-handling robots. This kit includes all the necessary reagents for the number of preps listed and requires the use of a magnetic accessory. Please see the Accessories Chapter (Chapter 6) to select the best magnetic accessory for your needs.

• Sample size: 25–50 μL
• High recovery of DNA fragments
• Rapid, one tube, 5 minute protocol

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

High-throughput Compatible
PCR Clean-Up Kit
5 min
DNA from Enzymatic Reactions (e.g. PCR)
ChargeSwitch™ Chemistry
PCR Products
Kit
ChargeSwitch™
960 Preps
Room Temperature
Next-Generation Sequencing (NGS), Cloning, Standard PCR
960 Preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.