missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix

Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.

Brand:  Invitrogen™ 11791100

840.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10033092

  • 1500.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
  • It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
  • Guarenteed stable for 6 months when properly stored at -20°C or -80°C

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
LR Clonase
100 Reactions
Clonase™, Gateway™
Dry Ice
Enzyme Buffer
LR Clonase Enzyme Mix
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.