missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix

Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.

Brand:  Invitrogen™ 11791020

348.65 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.



 View more versions of this product

Product Code. 10134992

  • 374.00 € / 20 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
  • It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
  • Guarenteed stable for 6 months when properly stored at -20°C or -80°C

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
LR Clonase
20 Reactions
Clonase™, Gateway™
Dry Ice
Enzyme Buffer
LR Clonase Enzyme Mix
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gateway LR Clonase II Enzyme mix

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H372-Causes damage to organs.
  • H361-Suspected of damaging fertility or the unborn child.
  • H351-Suspected of causing cancer.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P362-Take off contaminated clothing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Glycerin, 1, 4-Butanediamine, N-(3-aminopropyl)-, Proteinase, tritirachium albumserine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.