missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Polymerase I

DNA polymerase with 5'→3' and 3'→5' exodeoxyribonuclease activities

Brand:  Invitrogen™ 18010017

 View more versions of this product

Product Code. 10358002

  • 281.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 1000 units of DNA polymerase in a storage buffer of 50mM potassium phosphate (pH 7.0), 100mM KCl, 1mM DTT, and 50% (v/v) glycerol. It is supplied at 5-10 units/μL.
This item is not returnable. View return policy

Description

Description

Performance and quality testing
Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50mM potassium phosphate (pH 7.0), 6.7mM MgCl2 , 1mM 2-mercaptoethanol, 80μg/mL template/primer, 32μM dTTP, 69nM [3H]dTTP, and enzyme in 100μL for 30 min at 37°C. Also incorporates biotinylated nucleotides.

  • Applications: DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5' overhangs
  • Source: purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Cloning, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Liquid
3' - 5', 5' - 3'
Wet Ice
DNA Polymerase
DNA Polymerase I
250 U
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ DNA Polymerase I > 250 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.