missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment

DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities

Brand:  Invitrogen™ 18012021

 View more versions of this product

Product Code. 10388382

  • 182.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500mM Tris-HCl (pH 8.0), 100mM MgCl2, 500mM NaCl], vial of dilution buffer.
This item is not returnable. View return policy

Description

Description

Applications:
Fill-in of 5' overhangs . Synthesis of probes by random primers labeling method. Sequencing single- and double-stranded DNA. Site-directed mutagenesis.

Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.

Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.

Unit Definition:
One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.

Array CGH, Cloning, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Restriction Enzyme Cloning

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM
MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C.
DNA Polymerase I
100 Units
No
DNA Polymerase
3' to 5'
Approved for shipment on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.