Learn More
Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment
DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities
182.00 € - 670.00 €
Specifications
Content And Storage | DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C. |
---|---|
Polymerase | DNA Polymerase I |
Hot Start | No |
Includes: DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500mM Tris-HCl (pH 8.0), 100mM MgCl2, 500mM NaCl], vial of dilution buffer.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10388382
|
Invitrogen™
18012021 |
100 Units |
182.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10247092
|
Invitrogen™
18012039 |
500 Units |
670.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applications:
Fill-in of 5' overhangs . Synthesis of probes by random primers labeling method. Sequencing single- and double-stranded DNA. Site-directed mutagenesis.
Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.
Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.
Unit Definition:
One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.
Array CGH, Cloning, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Restriction Enzyme Cloning
Order Info
Shipping Condition: Approved for shipment on Wet or Dry Ice
Specifications
DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C. |
|
DNA Polymerase I | |
No |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.