missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human EpCAM Isolation Reagent (from cell culture)

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human EpCAM Isolation Reagent (from cell culture).

Brand:  Invitrogen™ 10618D

459.00 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 15470544

  • 540.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human EpCAM Isolation Reagent (from cell culture). Following isolation, the specific sub-population of exosomes, ECVs can be further studied using methods such as Western blotting, qPCR, or next generation sequencing.

  • Minimal hands-on time for specific exosome isolation
  • Compatible with most downstream analysis
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively isolated and studied
  • Cell culture media must first be enriched for exosomes
  • Can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation
  • Following enrichment, specific exosomes are isolated by binding to surface of magnetic beads
  • In addition to their high-quality sensitivity, reproducibility, and stability, Dynabeads allows you to see samples during handling due to color of magnetic beads
  • When a sample tube is placed in the magnet, bead-bound exosomes are pulled to side of tube, allowing for easy separation and buffer changes
  • Sample and bead volume can be easily scaled according to your sample size or downstream applications

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
Dynabeads™
Cell Cultures
Isolation Reagent
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2674852 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10618D
Lot Number2850829 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number10618D
Lot Number2893338 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number10618D
Lot Number3102574 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number10618D
Lot Number2974360 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number10618D
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Exosome-Human EpCAM Isolation Reagent (from cell culture) > 2mL; EpCAM isolation

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.