missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome Immunoprecipitation Reagent (Protein A)

Exosome Immunoprecipitation Reagent (Protein A) enables gentle magnetic isolation of exosomal proteins, causing minimal physical stress to the target protein.

Brand:  Invitrogen™ 10610D

161.50 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 13305364

  • 190.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Exosome Immunoprecipitation Reagent (Protein A) enables gentle magnetic isolation of exosomal proteins, causing minimal physical stress to the target protein.

  • Maintain intact exosome protein complexes
  • Reduce background significantly—low non-specific binding
  • Fast protocol time—only 30 minutes
  • Maximal comparison—ultra-sensitivity allows many samples on the same gel
  • Dynabeads coupled to Protein A are widely used for immunoprecipitation (IP), chromatin immunoprecipitation (ChIP), and protein isolation
  • The magnetic properties of Dynabeads makes them a superior alternative to sepharose or agarose slurry for immunoprecipitation, since magnetic separation technology is faster and gentler than other methods, causing minimal physical stress to the target proteins
  • Native protein conformation and intact, large protein complexes will be preserved
  • The binding capacity is approximately 240 μg human Ig per mL beads
  • For isolation of Ig via protein G, use Exosome Immunoprecipitation (Protein G)

Not high-throughput compatible.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2°C to 8°C.
Protein Complexes, Protein (Native)
Not High-throughput Compatible (Manual)
Protein A
1 mL
Room Temperature
2.8 μm
Exosome Immunoprecipitation Reagent
Beads in suspension
Magnetic Bead
Dynabeads™
Exosomes
All species
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Exosome Immunoprecipitation Reagent (Protein A) > 1mL; Protein A reagent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.