missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Streptavidin Isolation/Detection Reagent

Exosome-Streptavidin Isolation/Detection Reagent, when combined with your choice of biotinylated primary antibody, enables the purification of exosomes that can then be detected using techniques such as flow cytometry, electron microscopy, or Western blotting.

Brand:  Invitrogen™ 10608D

408.85 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 15256985

  • 481.00 € / 3mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Exosome-Streptavidin Isolation/Detection Reagent, when combined with your choice of biotinylated primary antibody, enables the purification of exosomes (also called extracellular vesicles and multi-vesicular bodies) that can then be detected using techniques such as flow cytometry, electron microscopy, or Western blotting. The exosomes must be pre-enriched before isolation. This can be done by ultracentrifugation, or by using the fast and efficient Total Exosome Isolation (from cell culture media) reagent.

  • Maximum flexibility with your choice of biotinylated antibody
  • See your sample during handling
  • Easily scalable protocol
  • Detect exosomes via flow cytometry, in less than 1 hour
  • Utilizes Dynabeads magnetic separation technology
  • Allows for easy purification of pre-enriched exosomes from cell culture media and then move on to detect the purified exosomes via techniques such as flow cytometry, electron microscopy, or Western blotting
  • Monodispersed and relatively large Dynabeads (4.5 μm in diameter) allow for a clear and defined FFC/SSC typically in less than 1 hour
  • Magnetic handling also allows sample to be visualized due to the light brown color of beads

Good Mixing is Critical

  • For successful exosome isolatio,n it is important to use a mixer that tilts and rotates to ensure that beads do not settle in tube
  • Avoid end-over end rotation for small sample volumes (e.g., 100 μL)
  • Please see user manual below for more guidelines
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cells in culture
Isolation/Detection Reagent
3 mL
Human
Store at 2°C to 8°C.
Dynabeads™
Cell Culture Supernatants
4.5 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2815219 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10608D
Lot Number2777734 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10608D
Lot Number2836955 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10608D
Lot Number2859260 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10608D
Lot Number2859263 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number10608D
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Exosome-Streptavidin Isolation/Detection Reagent > 3mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.