missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Untouched™ Human CD8 T Cells Kit

The Dynabeads™ Untouched™ Human CD8 T Cells Kit is used to isolate pure and viable untouched CD8+ T cells from PBMC by negative isolation.

Brand:  Invitrogen™ 11348D

946.90 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10268813

  • 1114.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: 10mL Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards human CD4, CD14, CD16a, CD16b, CD19, CD36, CD56, CD123 and CD235a (Glycophorin A)
This item is not returnable. View return policy

Description

Description

The Dynabeads™ Untouched™ Human CD8 T Cells Kit is used to isolate pure and viable untouched CD8+ T cells from PBMC by negative isolation. The kit depletes CD4+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes, and erythrocytes. The negatively isolated human CD8+ T-cells are left in the sample and have not been in contact with the Dynabeads™. The bead- and antibody-free CD8+ T cells are truly untouched, and in perfect shape for any functional assay or application.

Advantages of the Dynabeads™ Untouched™ Human CD8 T Cells Kit:
• High purity, recovery, and viability of human untouched CD8+ T cells
• Easy to use and to scale up
• No columns required

Magnetic bead-based Separation offers easy handling
An antibody mix towards the non-CD8 cells is added to the sample and allowed to bind to the cells. Dynabeads™ are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated and untouched human CD8+ T cells can be directly analyzed in a flow cytometer and used in any application. For depletion or positive isolation of CD8+ cells, select from our range of Human CD8+ T Cell Isolation Products.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

T Cells (CD8)
The Kit Contains: 10 mL Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards human CD4, CD14, CD16a, CD16b, CD19, CD36, CD56, CD123 and CD235a (Glycophorin A).

Storage Conditions: 2°C to 8°C. Do not freeze.
Cells
Negative Isolation
100 Isolations
DYNAL™, Dynabeads™, Untouched™
1 kit
Room Temperature
Human
Monoclonal
Cell Isolation Kit
Not High-throughput Compatible (Manual)
Processes ∼1 x 109 cells total
>95%, >98%
Research Grade
PBMC
1 x 107 PBMCs per isolation
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2809094 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11348D
Lot Number2801755 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11348D
Lot Number2813359 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11348D
Lot Number2801768 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11348D
Lot Number2825560 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11348D
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Dynabeads™ Untouched™ Human CD8 T Cells Kit > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.