Learn More
Multi Ion anion IC standard solution in H2O, Specpure™, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041693.AE
288.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Multi Ion anion IC standard solution in H2O is utilized as a standard solution in analytical chemistry and ion chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Multi Ion anion IC standard solution in H2O | |
100 mL | |
Liquid |
Br-, Cl-, F-, NO3-, PO4-3 and SO4-2at 100μg/mL. Keep refrigerated | |
MFCD00144814 | |
Miscible with water |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1539569 | Certificate of Analysis | 11/03/2025 | 041693.AE |
1525315 | Certificate of Analysis | 18/02/2025 | 041693.AE |
1517349 | Certificate of Analysis | 15/01/2025 | 041693.AE |
1510709 | Certificate of Analysis | 20/12/2024 | 041693.AE |
1499458 | Certificate of Analysis | 10/10/2024 | 041693.AE |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only