Learn More
Invitrogen™ Flp-In™ T-REx™ 293 Cell Line
Cells contain single stably integrated FRT site at transcriptionally active genomic locus
Brand: Invitrogen™ R78007
2992.00 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: 1 x 107 cells are supplied frozen in 1mL of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen.
Description
Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Targeted Integration

Specifications
1 vial of 1 x 107 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored. | |
Human | |
1 mL |
Flp-In T-Rex 293 | |
Flp-In™, T-REx™ |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.