missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ High Five™ Cells in Express Five™ Medium

Useful for the expression of recombinant proteins using the Baculovirus Expression Vector System (BEVS)

Brand:  Invitrogen™ B85502

1447.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10747474

  • 1608.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Review the Material Safety Data Sheet before handling.


Includes: High Five frozen cells, Express Five Serum-free Medium, Glutamine, 100X
This item is not returnable. View return policy

Description

Description

High Five Cells (BTI-TN-5B1-4) are a clonal isolate derived from the parental Trichopulsia ni cell line (cabbage looper ovary). High Five Insect Cells are adapted to serum-free culture in Express FiveSFM for maximum cell growth and recombinant gene expression.

  • Recombinant protein expression from a variety of expression systems
  • Up to ten-fold increase in secreted protein expression compared to Sf9 cells
  • Good growth in adherent or suspension culture
  • Quality and performance testing

Recombinant protein expression from a variety of expression systems High levels of protein expression in High Five cells can be obtained using either the BaculoDirect™ Baculovirus Expression System, the Bac-to-Bac™ Baculovirus Expression System, or the InsectDirect™ Expression System. Up to ten-fold increase in secreted protein expression compared to Sf9 cells High Five cells are the best choice for expression of secreted protein in insect cells. They consistently express 5–10 times more protein that Sf9 cells. Good growth in adherent or suspension culture High Five ™ cells thaw into adherent culture, forming an irregular monolayer with plaques that may be difficult to isolate. Protocols for adherent growth can be found in Express FiveSFM are available in the product manual. High Five cells can be adapted to suspension culture. Quality and performance testing Each lot of High Five cells is tested for cell growth and viability post-recovery from cryopreservation. In addition, the Master Seed Bank has been tested for contamination of bacteria, yeast, mycoplasma and virus and has been characterized by isozyme and karyotype analysis.

Bioproduction, Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Vaccine Production

TRUSTED_SUSTAINABILITY
Specifications

Specifications

High Five Frozen Cells:
• Storage conditions: liquid nitrogen (vapor phase)

Express Five Serum-free Medium:
• Storage conditions: 2°–8°C
• Protect from light

L-glutamine, 100X:
• Storage conditions: -5 to -20°C
• Protect from light
Insect Cells
Express Five™ SFM (Serum-Free Media)
High Five™
3 x 106 cells
High Five
Trichoplusia ni
3 x 106 cells
High Five Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ High Five™ Cells in Express Five™ Medium > 3 x 106 cells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.