missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K Solution

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Brand:  Invitrogen™ 4333793

Product Code. 15314118

  • 236.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range (optimal activity between 6.5 and 9.5)
  • Under denaturing conditions (e.g., the presence of SDS or urea)
  • In the presence of metal chelating agents (e.g. EDTA)
  • At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and Quality Testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit Definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNase-Free, RNase-Free
Store in refrigerator (2°C to 8°C).
5 mL
Liquid
Chromatin Biology/RNA Extraction/DNA Extraction
Wet Ice
Proteinase K
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2312002 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4333793
Lot Number2411005 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number4333793
Lot Number2308003 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog Number4333793
Lot Number2304002 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number4333793
Lot Number2303001 Certificate TypeCertificate of Analysis Date06/05/2023 Catalog Number4333793
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Proteinase K Solution > 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.